Merck
  • Linoleic acid metabolite drives severe asthma by causing airway epithelial injury.
Linoleic acid metabolite drives severe asthma by causing airway epithelial injury.
Scientific reports (2013-02-28)
Ulaganathan Mabalirajan, Rakhshinda Rehman, Tanveer Ahmad, Sarvesh Kumar, Suchita Singh, Geeta D Leishangthem, Jyotirmoi Aich, Manish Kumar, Kritika Khanna, Vijay P Singh, Amit K Dinda, Shyam Biswal, Anurag Agrawal, Balaram Ghosh
ABSTRACT
Airway epithelial injury is the hallmark of various respiratory diseases, but its mechanisms remain poorly understood. While 13-S-hydroxyoctadecadienoic acid (13-S-HODE) is produced in high concentration during mitochondrial degradation in reticulocytes little is known about its role in asthma pathogenesis. Here, we show that extracellular 13-S-HODE induces mitochondrial dysfunction and airway epithelial apoptosis. This is associated with features of severe airway obstruction, lung remodeling, increase in epithelial stress related proinflammatory cytokines and drastic airway neutrophilia in mouse. Further, 13-S-HODE induced features are attenuated by inhibiting Transient Receptor Potential Cation Channel, Vanilloid-type 1 (TRPV1) both in mouse model and human bronchial epithelial cells. These findings are relevant to human asthma, as 13-S-HODE levels are increased in human asthmatic airways. Blocking of 13-S-HODE activity or disruption of TRPV1 activity attenuated airway injury and asthma mimicking features in murine allergic airway inflammation. These findings indicate that 13-S-HODE induces mitochondrial dysfunction and airway epithelial injury.
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Linoleic acid sodium salt, ≥97% (GC)
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)