Merck
Guidance of cell migration by substrate dimension.
Biophysical journal (2013-02-28)
Stephanie S Chang, Wei-hui Guo, Youngeun Kim, Yu-li Wang
ABSTRACT
There is increasing evidence to suggest that physical parameters, including substrate rigidity, topography, and cell geometry, play an important role in cell migration. As there are significant differences in cell behavior when cultured in 1D, 2D, or 3D environments, we hypothesize that migrating cells are also able to sense the dimension of the environment as a guidance cue. NIH 3T3 fibroblasts were cultured on micropatterned substrates where the path of migration alternates between 1D lines and 2D rectangles. We found that 3T3 cells had a clear preference to stay on 2D rather than 1D substrates. Cells on 2D surfaces generated stronger traction stress than did those on 1D surfaces, but inhibition of myosin II caused cells to lose their sensitivity to substrate dimension, suggesting that myosin-II-dependent traction forces are the determining factor for dimension sensing. Furthermore, oncogene-transformed fibroblasts are defective in mechanosensing while generating similar traction forces on 1D and 2D surfaces. Dimension sensing may be involved in guiding cell migration for both physiological functions and tissue engineering, and for maintaining normal cells in their home tissue.
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 750, contains 900-1100 ppm MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 550, contains 80-120 ppm MEHQ as inhibitor, 270-330 ppm BHT as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 2000, contains ~1000 ppm MeHQ as stabilizer
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 20,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 10,000, contains MEHQ as inhibitor
Sigma-Aldrich
Poly(ethylene glycol) dimethacrylate, average Mn 6,000, contains 1000 ppm 4-methoxyphenol as inhibitor
Sigma-Aldrich
Polyacrylamide, nonionic water-soluble polymer
Sigma-Aldrich
Polyacrylamide, average Mn 150,000