Merck
  • Role of ERK1/2 kinase in the expression of iNOS by NDMA in human neutrophils.
Role of ERK1/2 kinase in the expression of iNOS by NDMA in human neutrophils.
Indian journal of experimental biology (2013-02-28)
Wioletta Ratajczak-Wrona, Ewa Jablonska, Marzena Garley, Jakub Jablonski, Piotr Radziwon
ABSTRACT
Potential role of ERK1/2 kinase in conjunction with p38 in the regulation of inducible nitric oxide synthase (iNOS) expression and nitric oxide (NO) production, and superoxide anion generation by human neutrophils (PMNs) exposed to N-nitrosodimethylamine (NDMA) was determined. Increased synthesis of NO due to the involvement of iNOS in neutrophils exposed to NDMA was observed. In addition, intensified activation of ERK1/2 and p38 kinases was determined in these cells. Inhibition of kinase regulated by extracellular signals (ERK1/2) pathway, in contrast to p38 pathway, led to an increased production of NO and expression of iNOS in PMNs. Moreover, as a result of inhibition of ERK1/2 pathway, a decreased activation of p38 kinase was observed in neutrophils, while inhibition of p38 kinase did not affect activation of ERK1/2 pathway in these cells. An increased ability to release superoxide anion by the studied PMNs was observed, which decreased after ERK1/2 pathway inhibition. In conclusion, in human neutrophils, ERK1/2 kinase is not directly involved in the regulation of iNOS and NO production induced by NDMA; however, the kinase participates in superoxide anion production in these cells.
Product Description
Supelco
N-Nitrosodimethylamine (NDMA) solution, certified reference material, 200 μg/mL in methanol