Merck
  • Quantum dots-based fluorescent probes for turn-on and turn-off sensing of butyrylcholinesterase.
Quantum dots-based fluorescent probes for turn-on and turn-off sensing of butyrylcholinesterase.
Biosensors & bioelectronics (2013-02-23)
Zhenzhen Chen, Xiangling Ren, Xianwei Meng, Longfei Tan, Dong Chen, Fangqiong Tang
ABSTRACT
A novel turn-on and turn-off sensor based on the fluorescence change of quantum dots (QDs) has been developed to detect the activity of butyrylcholinesterase (BChE). In the turn-on sensing system, we realized the detection of BChE with just one enzyme. A linear calibration plot of the activity of BChE was obtained in the wide amounts range from 10 to 1000 U/L and the detection limit was 10 U/L. In the turn-off sensing system, we realized the sensing of BChE with a wide linear relationship of 10-2000 U/L which was much wider than many other detection methods. We also studied the application in serum sample detection. The BChE sensor shows great performances. Furthermore, the turn-on and turn-off sensing mechanisms were studied extensively. These results showed that our strategy would most probably be applicable in assembling diagnostic micro-device for realizing the rapid clinical analysis of BChE.
Sigma-Aldrich
Thioglycolic acid solution, ~70 % (w/w) in H2O
Sigma-Aldrich
2-Mercaptopropionic acid, ≥95%, FG
Supelco
Thioglycolic acid solution, ~80% in H2O, for spectrophotometric det. of palladium, iron, uranium(VI), molybdates and nitrites
Sigma-Aldrich
Sodium thioglycolate, ≥96.5% (iodometric)
Sigma-Aldrich
Thiolactic acid, 95%
Sigma-Aldrich
Thioglycolic acid, ≥99%
Sigma-Aldrich
Thioglycolic acid, 98%