Merck
  • Monitoring of chronic Cannabis abuse: an LC-MS/MS method for hair analysis.
Monitoring of chronic Cannabis abuse: an LC-MS/MS method for hair analysis.
Journal of pharmaceutical and biomedical analysis (2013-01-12)
Laura Mercolini, Roberto Mandrioli, Michele Protti, Matteo Conti, Giovanni Serpelloni, Maria Augusta Raggi
ABSTRACT
An advanced analytical method based on liquid chromatography-tandem mass spectrometry (LC-MS/MS), has been developed for the identification and determination in hair of Δ(9)-tetrahydrocannabinol together with its major metabolite 11-nor-9-carboxy-Δ(9)-tetrahydrocannabinol. Since the latter is formed endogenously, it allows the assessment of chronic use excluding passive exposure to Cannabis. The sample pre-treatment procedure is based on a feasible incubative extraction followed by a liquid-liquid extraction step. Chromatographic separation was performed using a reversed-phase column and gradient elution with a formic acid/acetonitrile/water mobile phase. The limits of quantitation and of detection were 3pg/mg and 1pg/mg, respectively, for both analytes. The method was successfully applied to the analysis of hair samples from Cannabis abusers; the analyte concentrations found ranged from 55 to 100pg/mg for Δ(9)-tetrahydrocannabinol and from 5 to 10pg/mg for 11-nor-9-carboxy-Δ(9)-tetrahydrocannabinol. Accuracy studies also gave satisfactory results (recovery>87%), thus confirming the suitability of the assay for chronic consumption monitoring.
Product Description
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-11-nor-9-Carboxy-Δ9-THC solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-trans-Δ9-THC solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Δ9-Tetrahydrocannabinol solution, 1.0 mg/mL in methanol, analytical standard, for drug analysis