Merck
  • Evaluation of ceftiofur crystalline free acid sterile suspension for control of metritis in high-risk lactating dairy cows.
Evaluation of ceftiofur crystalline free acid sterile suspension for control of metritis in high-risk lactating dairy cows.
Theriogenology (2013-01-08)
C L McLaughlin, E P Stanisiewski, C A Risco, J E P Santos, G E Dahl, R C Chebel, C LaGrow, C Daugherty, L Bryson, D Weigel, J Hallberg, M J Lucas
ABSTRACT
Two studies were conducted to determine if clinical metritis could be prevented or decreased in at-risk lactating dairy cows by a single treatment with Excede Sterile Suspension (ceftiofur crystalline free acid sterile suspension [CCFA-SS]) administered within 24 hours after an abnormal calving. Study 1 was a preliminary study and study 2 was a clinical trial (designed to confirm the results of study 1). In both studies, abnormal calving was defined as cows that had dystocia (required assistance), twins, abortion, retained fetal membranes for 12 hours or more, or any combination thereof. A randomized block design with cows blocked on order-of-entry within dairy without regard to parity was used in both studies. In study 1, cows that had abnormal calving from six commercial dairies were randomly assigned to either untreated control (N = 122) or 6.6 mg ceftiofur equivalents/kg of body weight sc in the base of the ear (CCFA-SS, N = 121), within 24 hours after calving. Cows with normal calving during the enrollment period received no treatment and were included for observational purposes (N = 122). Health observations and rectal temperatures were recorded daily, and physical examinations were conducted on Days 1 ± 1, 7 ± 2, 14 ± 2, and 21 ± 2, and uterine swabs (for bacterial culture) were collected from a subsample of cows on Days 3 or 4, 7 ± 2, 14 ± 2, and 21 ± 2. These observations were made by treatment-blinded personnel. In study 2, cows with abnormal calving from 12 commercial dairies were assigned to receive either saline (control, N = 247) or CCFA-SS (N = 247) within 24 hours after calving. Health observations and rectal temperatures were recorded daily, and physical examinations were conducted on Days 0 to 2, 7 ± 1, and 14. In study 1, the incidence of metritis on Day 14 ± 2 was 20.2% versus 36.8% for CCFA-SS and control, respectively, with an odds ratio of 2.30 (P < 0.05). In study 2, incidences of metritis on Day 14 were 28.7% versus 43.5% for CCFA-SS and saline, respectively, with an odds ratio of 1.92 (P < 0.05). Rectal temperatures on Days 1 and 2 and the average for the first 6 days were lower (P < 0.05) for CCFA-SS compared with control cows for both studies. Treatment of cows with a single dose of CCFA-SS within 24 hours after abnormal calving reduced the incidence of subsequent metritis in lactating dairy cows.
Product Description
Supelco
Ceftiofur hydrochloride, VETRANAL®, analytical standard