Merck
Synthesis of tetrapeptide p-nitroanilides catalyzed by pepsin.
International journal of peptide and protein research (1993-02-01)
C A Abdel Malak, G I Lavrenova, E N Lysogorskaya, Filippova IYu, Terent'eva EYu, V M Stepanov
ABSTRACT
Swine pepsin at pH 5 efficiently catalyzes a condensation between Z-Ala-Ala-Phe-OH and p-nitroanilides of Leu, Phe, Val, Ala and Arg that leads to formation of corresponding benzyloxycarbonyl-tetrapeptide p-nitroanilides with yields of 70-90%. These reactions are complicated by co-precipitation of pepsin and the reaction products that necessitates the use of a relatively high concentration of pepsin.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg