Merck
  • Cations in a molecular funnel: vibrational spectroscopy of isolated cyclodextrin complexes with alkali metals.
Cations in a molecular funnel: vibrational spectroscopy of isolated cyclodextrin complexes with alkali metals.
Chemphyschem : a European journal of chemical physics and physical chemistry (2012-12-15)
Francisco Gámez, Paola Hurtado, Ana R Hortal, Bruno Martínez-Haya, Giel Berden, Jos Oomens
ABSTRACT
The benchmark inclusion complexes formed by α-cyclodextrin (αCD) with alkali-metal cations are investigated under isolated conditions in the gas phase. The relative αCD-M(+) (M=Li(+), Na(+), K(+), Cs(+)) binding affinities and the structure of the complexes are determined from a combination of mass spectrometry, infrared action spectroscopy and quantum chemical computations. Solvent-free laser desorption measurements reveal a trend of decreasing stability of the isolated complexes with increasing size of the cation guest. The experimental infrared spectra are qualitatively similar for the complexes with the four cations investigated, and are consistent with the binding of the cation within the primary face of the cyclodextrin, as predicted by the quantum computations (B3LYP/6-31+G*). The inclusion of the quantum-chemical cation disrupts the C(6) symmetry of the free cyclodextrin to provide the optimum coordination of the cations with the -CH(2)OH groups in C(1), C(2) or C(3) symmetry arrangements that are determined by the size of the cation.
Sigma-Aldrich
α-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
α-Cyclodextrin, ≥98%
Sigma-Aldrich
α-Cyclodextrin, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.0-101.0% cyclodextrin basis (HPLC)
Sigma-Aldrich
α-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (HPLC)
Sigma-Aldrich
α-Cyclodextrin, purum, ≥98.0% (HPLC)