Merck
  • Aqueous levels of fluocinolone acetonide after administration of fluocinolone acetonide inserts or fluocinolone acetonide implants.
Aqueous levels of fluocinolone acetonide after administration of fluocinolone acetonide inserts or fluocinolone acetonide implants.
Ophthalmology (2012-12-12)
Peter A Campochiaro, Quan Dong Nguyen, Gulnar Hafiz, Steven Bloom, David M Brown, Miguel Busquets, Thomas Ciulla, Leonard Feiner, Nelson Sabates, Kathleen Billman, Barry Kapik, Ken Green, Frances E Kane
ABSTRACT
To compare aqueous levels of fluocinolone acetonide (FAc) after administration of FAc inserts or FAc implants (Retisert; Bausch & Lomb, Rochester, NY). Comparison of pharmacokinetics from 2 prospective, interventional, clinical trials. Thirty-seven patients with diabetic macular edema (DME) (Fluocinolone Acetonide in Human Aqueous [FAMOUS] Study, C-01-06-002) and 7 patients with uveitis (NA-00019318). Aqueous FAc was measured after administration of FAc implants or 0.2 μg/day (low dose, ILUVIEN; Alimera Sciences Inc., Alpharetta, GA) or 0.5 μg/day (high dose) FAc inserts. The primary end point was aqueous levels of FAc. At 1 month after administration for subjects who received 1 treatment, mean aqueous FAc levels were 2.17 (low dose) and 3.03 ng/ml (high dose) for FAc inserts and 6.12 ng/ml for FAc implants with maximum levels of 3.83, 6.66, and 13.50 ng/ml, respectively. At 3 months, mean FAc levels were 1.76, 2.15, and 6.12 ng/ml, respectively. Between 6 and 36 months after low-dose inserts, aqueous levels of FAc were remarkably stable, ranging from 1.18 to 0.45 ng/ml. After high-dose inserts, mean FAc levels were stable between 6 and 24 months, ranging from 1.50 to 0.84 ng/ml and then decreasing to 0.35 ng/ml at 30 months and 0.15 ng/ml at 36 months. In implant-containing eyes, mean FAc levels remained >6 ng/ml through 15 months, the last time point with measurements from at least 6 eyes. Low- and high-dose FAc inserts both provide stable long-term release of FAc with comparable peak levels in the aqueous: slightly >2 ng/ml for approximately 3 months followed by steady-state levels between 1.0 and 0.5 ng/ml through 36 months for low-dose inserts versus levels between 1.5 and 1.1 ng/ml through 24 months for high-dose inserts. Steady-state aqueous levels after FAc implants were >6 ng/ml. These results provide new insights that aid in the interpretation of efficacy trials and indicate that there is a dose effect for steroid-induced ocular hypertension. In susceptible patients, prolonged aqueous levels of FAc >1 ng/ml moderately increased the risk of glaucoma and levels >6 ng/ml posed a markedly increase risk.
Product Description
Fluocinolone acetonide, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluocinolone acetonide, analytical standard