Merck
  • MRT letter: localization of endogenous hydrogen peroxide by modified processes of sample preparation for transmission electron microscope in Escherichia coli.
MRT letter: localization of endogenous hydrogen peroxide by modified processes of sample preparation for transmission electron microscope in Escherichia coli.
Microscopy research and technique (2012-11-20)
Xin Li, Rongliu Hu, Wenxue Zhu, Jinling Fan, Xinyue Pang, Na Wang, Liping Wang, Lipeng Yang, Chunyan Zhao, Chenyang He
ABSTRACT
The bacterial endogenous hydrogen peroxide (H(2)O(2)) was detected cytochemically by its reaction with cerium chloride (CeCl(3)) to produce electron-dense deposits of cerium perhydroxides. The sequence of fixation and CeCl(3) staining of H(2)O(2) in the processing of transmission electron microscope (TEM) sample preparation is crucial to the localization of endogenous H(2)O(2) in Escherichia coli. In this study, results confirmed that the process that fixation simultaneously with CeCl(3) staining provided optimum effects for H(2)O(2) localization in E. coli. The modified process of TEM provides very efficient protection for H(2)O(2) localization and more accurate quantization for the H(2)O(2) accumulation in bacterial cells.
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.9% trace metals basis
Sigma-Aldrich
Cerium(III) chloride heptahydrate, 99.999% trace metals basis
Sigma-Aldrich
Cerium(III) chloride heptahydrate, purum p.a., ≥98.0% (AT)
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Cerium(III) chloride, AnhydroBeads, −10 mesh, 99.9%