Merck
  • More than a marine propeller--the flagellum of the probiotic Escherichia coli strain Nissle 1917 is the major adhesin mediating binding to human mucus.
More than a marine propeller--the flagellum of the probiotic Escherichia coli strain Nissle 1917 is the major adhesin mediating binding to human mucus.
International journal of medical microbiology : IJMM (2012-11-08)
Anja Troge, Wolfgang Scheppach, Bjoern O Schroeder, Stefan A Rund, Klaus Heuner, Jan Wehkamp, Eduard F Stange, Tobias A Oelschlaeger
ABSTRACT
The flagellum of the probiotic Escherichia coli strain Nissle 1917 (EcN) is not just responsible for motility, but also for EcN's ability to induce the production of human β-defensin 2. Here, we report a third function of this EcN organell. In this study we investigated the role of the EcN flagellum in adhesion to different host tissues by ex vivo and in vitro studies. Ex vivo studies with cryosections of human gut biopsies revealed that the flagellum of EcN is most likely important for efficient adhesion to the human intestinal tract. These results and in vitro studies with different epithelial cells indicated that the presence of mucus is important for efficient mediation of adhesion by the flagellum of EcN. We observed direct interaction between isolated flagella from EcN wild type and porcine mucin 2 as well as human mucus. However, we could not observe any interaction of the flagella with murine mucus. For the first time, we identified the mucus component gluconate as one receptor for the binding of flagella from EcN and were able to exclude the flagellin domain D3 as a responsible interaction partner. We propose that the flagellum of EcN is its major adhesin in vivo, which enables this probiotic strain to compete efficiently for binding sites on host tissue with several bacterial pathogens.
Sigma-Aldrich
Sodium gluconate, meets USP testing specifications
Supelco
D-Gluconic acid sodium salt, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
D-Gluconic acid sodium salt, ≥99% (HPLC)
Sigma-Aldrich
D-Gluconic acid solution, 49-53 wt. % in H2O
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Iron(II) D-gluconate dihydrate, 98%
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Sigma-Aldrich
Magnesium D-gluconate hydrate, 98.0-102% anhydrous basis, meets USP testing specifications
Sigma-Aldrich
Magnesium D-gluconate hydrate, ≥98% (HPLC)
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)