Merck
  • Randomized trial of myeloablative conditioning regimens: busulfan plus cyclophosphamide versus busulfan plus fludarabine.
Randomized trial of myeloablative conditioning regimens: busulfan plus cyclophosphamide versus busulfan plus fludarabine.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology (2012-11-07)
Je-Hwan Lee, Young-Don Joo, Hawk Kim, Hun Mo Ryoo, Min Kyoung Kim, Gyeong-Won Lee, Jung-Hee Lee, Won-Sik Lee, Jae-Hoo Park, Sung-Hwa Bae, Myung Soo Hyun, Dae-Young Kim, Sung-Doo Kim, Young Joo Min, Kyoo-Hyung Lee
ABSTRACT
We conducted a phase III randomized clinical trial to compare two myeloablative conditioning regimens for allogeneic hematopoietic cell transplantation (HCT) in patients with leukemia and myelodysplastic syndrome. After randomization, 64 patients received busulfan (3.2 mg/kg per day × 4 days) plus cyclophosphamide (60 mg/kg per day × 2 days; BuCy), and 62 patients received busulfan (same dose and schedule) plus fludarabine (30 mg/m(2) per day × 5 days; BuFlu). The median age was 41 years (range, 17 to 59 years). Five patients in the BuFlu arm experienced graft failure (primary, n = 1; secondary, n = 4). At 4 weeks after HCT, the median percentage of recipient hematopoietic chimerism was significantly greater in the BuFlu arm (0% v 5.5%; P < .001), and complete donor chimerism was greater in the BuCy arm (97.2% v 44.4%; P < .001). Severe (grade 3 or higher) infection and gastrointestinal adverse events were significantly more common in the BuCy arm, but the frequencies of hepatic adverse events were similar in the two arms. Nonrelapse mortality was similar in the two arms, but the BuCy arm had better overall survival (OS), relapse-free survival (RFS), and event-free survival (EFS; OS at 2 years, 67.4% v 41.4%, P = .014; RFS, 74.7% v 54.9%, P = .027; EFS, 60.7% v 36.0%, P = .014). Our results indicate that the BuFlu regimen is not a suitable replacement for the BuCy regimen in young adults who are eligible for myeloablative conditioning therapy for allogeneic HCT.
Product Description
Busulfan, European Pharmacopoeia (EP) Reference Standard
Supelco
Busulfan, analytical standard, for drug analysis