Merck
  • Determination of six microcystins and nodularin in surface and drinking waters by on-line solid phase extraction-ultra high pressure liquid chromatography tandem mass spectrometry.
Determination of six microcystins and nodularin in surface and drinking waters by on-line solid phase extraction-ultra high pressure liquid chromatography tandem mass spectrometry.
Journal of chromatography. A (2012-10-31)
Eduardo Beltrán, María Ibáñez, Juan Vicente Sancho, Félix Hernández
ABSTRACT
Microcystins and nodularin are cyclic peptides hepatotoxins produced by cyanobacterial genera (blue-green algae). Toxic cyanobacterial blooms are a worldwide problem, as reported in several countries, like China, Australia, or the United States. Therefore, it is necessary to develop sensitive and reliable analytical methodology to determine this type of toxins in water at parts per billion levels, or even lower. In this work, the potential of solid-phase extraction coupled on-line to ultra-high-pressure liquid chromatography/electrospray tandem mass spectrometry (SPE-UHPLC-MS/MS) has been investigated for the efficient quantification and confirmation of microcystins LR, RR, YR, LY, LW, LF and nodularin in surface and drinking water samples, at sub-ppb levels. The method developed involves the injection of only 1 mL of water sample into the on-line SPE-UHPLC-MS/MS system and allows the rapid determination of the compounds selected (8 min of chromatographic run), avoiding laborious sample treatment. The method was validated in surface and drinking water by means of recovery experiments at 0.25 and 1 μg L(-1). Average recoveries (n=5) ranged from 71 to 116%, with relative standard deviations (RSDs) lower than 15%. For microcystins LR, RR, YR and nodularin, a third level was also assayed (0.1 μg L(-1)) obtaining satisfactory data too. Limits of detection between 0.002 and 0.0405 μg L(-1) were estimated (0.0005 μg L(-1) for nodularin). The developed method was applied to the analysis of water samples collected in the province of Castellón (Spain). The acquisition of three MS/MS transitions for each compound allowed the unequivocal confirmation of positive samples, which was supported by the accomplishment of ion intensity ratios and retention time when compared with reference standards.
Product Description
Supelco
Nodularin solution, 10 μg/mL in methanol, analytical standard