Merck
  • Catalytic ozonation of sulphamethoxazole in the presence of carbon materials: catalytic performance and reaction pathways.
Catalytic ozonation of sulphamethoxazole in the presence of carbon materials: catalytic performance and reaction pathways.
Journal of hazardous materials (2012-09-27)
Alexandra G Gonçalves, José J M Órfão, Manuel Fernando R Pereira
ABSTRACT
Two carbon materials (multi-walled carbon nanotubes, MWCNTs, and activated carbon) were investigated as ozonation catalysts for the mineralization of the antibiotic sulphamethoxazole (SMX). MWCNTs presented a higher catalytic performance than activated carbons, which was justified by their differences in surface chemistry and by the higher internal mass transfer resistances expected for activated carbons. 3-Amino-5-methylisoxazole and p-benzoquinone were detected as primary products of single and catalytic ozonation of SMX, whereas oxamic, oxalic, pyruvic and maleic acids were identified as refractory final oxidation products. The original sulphur of the SMX was almost completely converted to sulphate and part of the nitrogen was converted to NH4+ and NO3-. The presence of the radical scavenger tert-butanol during catalytic and single ozonation evidenced the participation of HO radicals in the oxidation mechanisms of SMX, especially in the mineralization of several intermediates. Microtox tests revealed that simultaneous use of ozone and MWCNTs originated lower acute toxicity. The time course of all detected compounds was studied and the transformation pathway for the complete mineralization of SMX by single and catalytic ozonation in the presence of the selected materials was elucidated.
Product Description
Supelco
Sulfamethoxazole
Supelco
Sulfamethoxazole, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Sulfamethoxazole, VETRANAL®, analytical standard
Supelco
Sulfamethoxazole, Pharmaceutical Secondary Standard; Certified Reference Material