Merck
  • Surface characterization and biocompatibility of titanium alloys implanted with nitrogen by Hardion+ technology.
Surface characterization and biocompatibility of titanium alloys implanted with nitrogen by Hardion+ technology.
Journal of materials science. Materials in medicine (2012-08-25)
D M Gordin, T Gloriant, V Chane-Pane, D Busardo, V Mitran, D Höche, C Vasilescu, S I Drob, A Cimpean
ABSTRACT
In this study, the new Hardion+ micro-implanter technology was used to modify surface properties of biomedical pure titanium (CP-Ti) and Ti-6Al-4V ELI alloy by implantation of nitrogen ions. This process is based on the use of an electron cyclotron resonance ion source to produce a multienergetic ion beam from multicharged ions. After implantation, surface analysis methods revealed the formation of titanium nitride (TiN) on the substrate surfaces. An increase in superficial hardness and a significant reduction of friction coefficient were observed for both materials when compared to non-implanted samples. Better corrosion resistance and a significant decrease in ion release rates were observed for N-implanted biomaterials due to the formation of the protective TiN layer on their surfaces. In vitro tests performed on human fetal osteoblasts indicated that the cytocompatibility of N-implanted CP-Ti and Ti-6Al-4V alloy was enhanced in comparison to that of the corresponding non treated samples. Consequently, Hardion+ implantation technique can provide titanium alloys with better qualities in terms of corrosion resistance, cell proliferation, adhesion and viability.
Sigma-Aldrich
Titanium nitride, <3 μm