Merck
  • Tris(hydroxymethyl) aminomethane salt of ramipril: synthesis, structural characterization from X-ray powder diffraction and stability studies.
Tris(hydroxymethyl) aminomethane salt of ramipril: synthesis, structural characterization from X-ray powder diffraction and stability studies.
Journal of pharmaceutical and biomedical analysis (2012-08-11)
Abir Bhattacharya, Basab Chattopadhyay, Santu Chakraborty, Bhairab N Roy, Girij P Singh, Himangshu M Godbole, Umesh B Rananaware, Alok K Mukherjee
ABSTRACT
Tris(hydroxymethyl) aminomethane (tris) salt of API ramipril was synthesized, and characterized by FTIR, TG-DSC and ab initio X-ray powder structure analysis. The compound, ramipril-tris (II), crystallizes in the monoclinic space group P2(1) with a=24.3341(15), b=6.4645(5), c=9.5357(7) Å, β=96.917(3)° and V=1489.1(3) Å(3). The crystal structure has been determined from laboratory X-ray powder diffraction data using direct space global optimization strategy (simulated annealing) followed by the Rietveld refinement. A network of intermolecular OH…O, CH…N and CH…O hydrogen bonds between the ramipril-ramipril, tris-tris and ramipril-tris components in the compound generates a two-dimensional molecular assembly in (110) plane. A comparative study of solid-state stabilities of ramipril-tris (II) with that of ramipril (I) and ramipril-erbumine (III) indicates that ramipril-tris (II) is the most stable one among the three, and the conversion to impurity D after 72 h at 80 °C is only 1.5%. The solution phase analysis at different pH values also reveals a greater stability of ramipril-tris (II) over ramipril (I).
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
SAFC
Tromethamine
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline