Merck
  • Effective and selective extraction of noble metal nanoparticles from environmental water through a noncovalent reversible reaction on an ionic exchange resin.
Effective and selective extraction of noble metal nanoparticles from environmental water through a noncovalent reversible reaction on an ionic exchange resin.
Chemical communications (Cambridge, England) (2012-08-07)
Lingxiangyu Li, Kerstin Leopold, Michael Schuster
ABSTRACT
Capable of preserving the size and shape of nanoparticles, a novel method to effectively and selectively extract noble metal nanoparticles even at the 80 ng L(-1) level from real environmental water was designed and performed using a noncovalent reversible adsorption onto an ionic exchange resin.
Sigma-Aldrich
Mercaptosuccinic acid, 97%
Sigma-Aldrich
Mercaptosuccinic acid, ReagentPlus®, ≥99.0% (HPLC)