Merck
  • Modified asymmetric Strecker reaction of aldehyde with secondary amine: a protocol for the synthesis of S-clopidogrel (an antiplatelet agent).
Modified asymmetric Strecker reaction of aldehyde with secondary amine: a protocol for the synthesis of S-clopidogrel (an antiplatelet agent).
The Journal of organic chemistry (2012-07-28)
Arghya Sadhukhan, S Saravanan, Noor-ul H Khan, Rukhsana I Kureshy, Sayed H R Abdi, Hari C Bajaj
ABSTRACT
A first approach for catalytic asymmetric Strecker reaction of aldehydes with a secondary amine in the presence of sodium fluoride using hydroquinine as chiral catalyst was developed. The catalytic system gave α-aminonitriles in excellent yields (up to 95%) and high enantioselectivities (er up to 94:6). The efficacy of the chiral product was successfully fulfilled in the improved synthesis of (S)-clopidogrel (an antiplatelet agent).
Sigma-Aldrich
Hydroquinidine hydrochloride, 98%
Sigma-Aldrich
Hydroquinine, 98%