Merck
  • A modular synthesis of dithiocarbamate pendant unnatural α-amino acids.
A modular synthesis of dithiocarbamate pendant unnatural α-amino acids.
Chemical communications (Cambridge, England) (2012-07-21)
Amit Saha, R B Nasir Baig, John Leazer, Rajender S Varma
ABSTRACT
Unnatural α-amino acids containing dithiocarbamate side chains were synthesized by a one-pot reaction of in situ generated dithiocarbamate anions with sulfamidates. A wide range of these anions participated in the highly regio- and stereo-selective ring opening of sulfamidates to produce the corresponding dithiocarbamate pendant α-amino acids in high yields.
Product Description
Supelco
Sulfamic acid, analytical standard (for acidimetry), ACS reagent
Sigma-Aldrich
Sulfamic acid, 99.999% trace metals basis
Sigma-Aldrich
Ammonium sulfamate, BioXtra, ≥98.0%
Sigma-Aldrich
Sulfamic acid, reagent grade, 98%
Sigma-Aldrich
Ammonium sulfamate, ACS reagent, ≥98.0%
Sigma-Aldrich
Sulfamic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfamic acid, ACS reagent, 99.3%