Merck
  • Characterization of bioimprinted tannase and its kinetic and thermodynamics properties in synthesis of propyl gallate by transesterification in anhydrous medium.
Characterization of bioimprinted tannase and its kinetic and thermodynamics properties in synthesis of propyl gallate by transesterification in anhydrous medium.
Applied biochemistry and biotechnology (2012-06-20)
Guangjun Nie, Zhiming Zheng, Guohong Gong, Genhai Zhao, Yan Liu, Junying Song, Jun Dai
ABSTRACT
Tannase has been extensively applied to synthesize gallic acid esters. Bioimprinting technique can evidently enhance transesterification-catalyzing performance of tannase. In order to promote the practical utilization of the modified tannase, a few enzymatic characteristics of the enzyme and its kinetic and thermodynamics properties in synthesis of propyl gallate by transesterification in anhydrous medium have been studied. The investigations of pH and temperature found that the imprinted tannase holds an optimum activity at pH 5.0 and 40 °C. On the other hand, the bioimprinting technique has a profound enhancing effect on the adapted tannase in substrate affinity and thermostability. The kinetic and thermodynamic analyses showed that the modified tannase has a longer half-time of 1,710 h at 40 °C; the kinetic constants, the activation energy of reversible thermal inactivation, and the activation energy of irreversible thermal inactivation, respectively, are 0.054 mM, 17.35 kJ mol(-1), and 85.54 kJ mol(-1) with tannic acid as a substrate at 40 °C; the free energy of Gibbs (ΔG) and enthalpy (ΔH) were found to be 97.1 and 82.9 kJ mol(-1) separately under the same conditions.
Sigma-Aldrich
Tannase from Aspergillus ficuum, powder, white, ≥150 U/g
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Propyl gallate, for microscopy, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, powder
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material