Merck
  • Ultrafast photo-induced charge transfer unveiled by two-dimensional electronic spectroscopy.
Ultrafast photo-induced charge transfer unveiled by two-dimensional electronic spectroscopy.
The Journal of chemical physics (2012-06-07)
Oliver Bixner, Vladimír Lukeš, Tomáš Mančal, Jürgen Hauer, Franz Milota, Michael Fischer, Igor Pugliesi, Maximilian Bradler, Walther Schmid, Eberhard Riedle, Harald F Kauffmann, Niklas Christensson
ABSTRACT
The interaction of exciton and charge transfer (CT) states plays a central role in photo-induced CT processes in chemistry, biology, and physics. In this work, we use a combination of two-dimensional electronic spectroscopy (2D-ES), pump-probe measurements, and quantum chemistry to investigate the ultrafast CT dynamics in a lutetium bisphthalocyanine dimer in different oxidation states. It is found that in the anionic form, the combination of strong CT-exciton interaction and electronic asymmetry induced by a counter-ion enables CT between the two macrocycles of the complex on a 30 fs timescale. Following optical excitation, a chain of electron and hole transfer steps gives rise to characteristic cross-peak dynamics in the electronic 2D spectra, and we monitor how the excited state charge density ultimately localizes on the macrocycle closest to the counter-ion within 100 fs. A comparison with the dynamics in the radical species further elucidates how CT states modulate the electronic structure and tune fs-reaction dynamics. Our experiments demonstrate the unique capability of 2D-ES in combination with other methods to decipher ultrafast CT dynamics.
Sigma-Aldrich
Tetrabutylammonium cyanide, technical, ≥80%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, technical, ~40% in H2O (~1.5 M)
Supelco
Tetrabutylammonium perchlorate, for electrochemical analysis, ≥99.0%
Supelco
Tetrabutylammonium hydroxide solution, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetrabutylammonium perchlorate, ≥95.0% (T)
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 53.5-56.5% in H2O
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic solution, 1.0 M in H2O
Sigma-Aldrich
Tetrabutylammonium cyanide, 95%
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 1.0 M in methanol
Sigma-Aldrich
Tetrabutylammonium hydroxide solution, 40 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium hydrogensulfate, 97%
Sigma-Aldrich
Tetrabutylammonium iodide, reagent grade, 98%
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Supelco
Tetrabutylammonium iodide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium phosphate monobasic, puriss., 99% (T)
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium bisulfate, puriss., ≥99.0% (T)
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Supelco
Tetrabutylammonium bisulfate solution, suitable for ion pair chromatography, LiChropur, concentrate, ampule
Sigma-Aldrich
Tetrabutylammonium iodide, ≥99.0% (AT)
Sigma-Aldrich
Tetrabutylammonium bisulfate solution, ~55% in H2O
Supelco
Tetrabutylammonium bisulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 1.0 M in THF
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium azide
Sigma-Aldrich
Tetrabutylammonium fluoride solution, 75 wt. % in H2O
Sigma-Aldrich
Tetrabutylammonium chloride, ≥97.0% (NT)
Sigma-Aldrich
Tetrabutylammonium nitrate, 97%
Supelco
Tetrabutylammonium chloride, suitable for ion pair chromatography, LiChropur, ≥99.0%