Merck
  • Separation and evaluation of soybean protein hydrolysates prepared by immobilized metal ion affinity chromatography with different metal ions.
Separation and evaluation of soybean protein hydrolysates prepared by immobilized metal ion affinity chromatography with different metal ions.
Journal of chromatographic science (2012-05-29)
He Liu, Xiao Lan Bao, Ying Lv, Jing Ting Xu, Shun Tang Guo
ABSTRACT
Metal ion affinity chromatography is widely used to purify peptides on the basis of the dissimilarities of their amino acids. However, researchers are interested in the separation differences between different metal ions in this method. In our study, four kinds of commonly used metal ions are compared by the amount of immobilized metal ion on iminodiacetic acid-Sepharose and binding amount of soybean peptide to immobilized iminodiacetic acid-Mn(+) adsorbents and evaluated by high-performance liquid chromatography (HPLC) profiles. The results show that due to the different adsorption behaviors of metal ions, the binding ability order of soybean protein peptide on the column should be Fe(3+) > Cu(2+) > Zn(2+) > Ca(2+). The HPLC profiles show that peptides adsorbed by four kinds of metal ions display similar strong hydrophobic characteristics.
Sigma-Aldrich
Iminodiacetic acid, 98%
Sigma-Aldrich
Iminodiacetic acid, purum, ≥98.0% (T)