Merck
  • Modifying the STM tip for the ' ultimate ' imaging of the Si(111)-7×7 surface and metal-supported molecules.
Modifying the STM tip for the ' ultimate ' imaging of the Si(111)-7×7 surface and metal-supported molecules.
Chimia (2012-05-02)
Ye-Liang Wang, Zhi-Hai Cheng, Zhi-Tao Deng, Hai-Ming Guo, Shi-Xuan Du, Hong-Jun Gao
ABSTRACT
We report on high-resolution STM measurements with modified probe tips. First, both the rest atoms and adatoms of a Si(111)-7×7 surface are observed simultaneously. The visibility of rest atoms is dependent upon the sample bias voltage (less than -0.7 V) and is enhanced by sharpening the tip, which is rationalized by first-principles calculations. Second, a tip with a perylene molecule adsorbed at its apex is used to discriminate the molecular states and the metal states of the underlying Ag(110) surface, which is attributable to a mismatch between the energy levels of the functionalized tip and the adsorbates on silver. Lastly, high-resolution images of iron phthalocyanine (FePc) and zinc phthalocyanine (ZnPc) molecules on Au(111) are obtained by using an O(2)-terminated tip, and the images reveal rich intramolecular features arising from molecular orbitals that are not observed when using clean metallic tips.
Sigma-Aldrich
Zinc phthalocyanine, Dye content 97 %
Pricing and availability is not currently available.