Merck
  • Dispersive microextraction based on water-coated Fe₃O₄ followed by gas chromatography-mass spectrometry for determination of 3-monochloropropane-1,2-diol in edible oils.
Dispersive microextraction based on water-coated Fe₃O₄ followed by gas chromatography-mass spectrometry for determination of 3-monochloropropane-1,2-diol in edible oils.
Journal of chromatography. A (2012-04-24)
Qin Zhao, Fang Wei, Neng Xiao, Qiong-Wei Yu, Bi-Feng Yuan, Yu-Qi Feng
ABSTRACT
In the present work, we developed a novel dispersive microextraction technique by combining the advantages of liquid-phase microextraction (LPME) and magnetic solid-phase extraction (MSPE). In this method, trace amount of water directly absorbed on bare Fe₃O₄ to form water-coated Fe₃O₄ (W-Fe₃O₄) and rapid extraction can be achieved while W-Fe₃O₄ dispersed in the sample solution. The analyte adsorbed W-Fe₃O₄ can be easily collected and isolated from sample solution by application of a magnet. It was worth noting that in the proposed method water was used as extractant and Fe₃O₄ served as the supporter and retriever of water. The performance of the method was evaluated by extraction of 3-monochloropropane-1,2-diol (3-MCPD) from edible oils. The extracted 3-MCPD was then derived by a silylanization reagent (1-trimethylsilylimidazole) before gas chromatography-mass spectrometry (GC-MS) analysis. Several parameters that affected the extraction and derivatization efficiency were investigated. Our results showed that the limit of detection for 3-MCPD was 1.1 ng/g. The recoveries in spiked oil samples were in the range of 70.0-104.9% with the RSDs less than 5.6% (intra-day) and 6.4% (inter-day). Taken together, the simple, rapid and cost-effective method developed in current study, offers a potential application for the extraction and preconcentration of hydrophilic analytes from complex fatty samples.
Sigma-Aldrich
1-(Trimethylsilyl)imidazole, 96%
Supelco
1-(Trimethylsilyl)imidazole, LiChropur
Sigma-Aldrich
1-(Trimethylsilyl)imidazole, ≥98.0%