Merck
  • [Improvement of 2-mercaptoimidazoline analysis in rubber products containing chlorine].
[Improvement of 2-mercaptoimidazoline analysis in rubber products containing chlorine].
Shokuhin eiseigaku zasshi. Journal of the Food Hygienic Society of Japan (2012-03-28)
Reiko Kaneko, Nahoko Haneishi, Yoko Kawamura
ABSTRACT
An improved analysis method for 2-mercaptoimidazoline in rubber products containing chlorine was developed. 2-Mercaptoimidazoline (20 µg/mL) is detected by means of TLC with two developing solvents in the official method. But, this method is not quantitative. Instead, we employed HPLC using water-methanol (9 : 1) as the mobile phase. This procedure decreased interfering peaks, and the quantitation limit was 2 µg/mL of standard solution. 2-Mercaptoimidazoline was confirmed by GC-MS (5 µg/mL) and LC/MS (1 µg/mL) in the scan mode. For preparation of test solution, a soaking extraction method, in which 20 mL of methanol was added to the sample and allowed to stand overnight at about 40°C, was used. This gave similar values to the Soxhlet extraction method (official method) and was more convenient. The results indicate that our procedure is suitable for analysis of 2-mercaptoimidazoline. When 2-mercaptoimidazoline is detected, it is confirmed by either GC/MS or LC/MS.
Sigma-Aldrich
2-Imidazolidinethione, purum, ≥98.0% (HPLC)
Sigma-Aldrich
2-Imidazolidinethione, 98%
Supelco
2-Imidazolidinethione, PESTANAL®, analytical standard