Merck
  • Oxidative and apoptotic effects of lambda-cyhalothrin modulated by piperonyl butoxide in the liver of Oreochromis niloticus.
Oxidative and apoptotic effects of lambda-cyhalothrin modulated by piperonyl butoxide in the liver of Oreochromis niloticus.
Environmental toxicology and pharmacology (2012-03-06)
Petek Piner, Nevin Uner
ABSTRACT
The aim of this study was to investigate the toxic effects of pyrethroid pesticide lambda-cyhalothrin in the presence of piperonyl butoxide as a modulator in the liver of juvenile Oreochromis niloticus. LC(50) (96h) value of lambda-cyhalothrin was determined as 2.901μg/L for O. niloticus. The fish were exposed to 0.48μg/L (1/6 of the 96-h LC(50)) lambda-cyhalothrin and 10μg/L piperonyl butoxide for 96-h and 15-d. tGSH, GSH, GSSG, Hsp70 and TBARS contents, GPx, GR, GST and caspase-3 enzymes activities were determined. Lambda-cyhalothrin caused increases in tGSH, GSH, TBARS contents, and GST activity. Piperonyl butoxide treatment with lambda-cyhalothrin caused significant increases in tGSH GSH, Hsp70, TBARS contents, and GPx and GST activities while caspase-3 activity was decreased. The results of the present study revealed that lambda-cyhalothrin caused oxidative stress which upregulated GSH and GSH-related enzymes. Piperonyl butoxide increased the oxidative stress potential and apoptotic effects of lambda-cyhalothrin.
Sigma-Aldrich
Piperonylbutoxide, technical grade, 90%
Supelco
Piperonylbutoxide, PESTANAL®, analytical standard