Merck
  • Design and synthesis of fluorescence-labeled closo-dodecaborate lipid: its liposome formation and in vivo imaging targeting of tumors for boron neutron capture therapy.
Design and synthesis of fluorescence-labeled closo-dodecaborate lipid: its liposome formation and in vivo imaging targeting of tumors for boron neutron capture therapy.
Organic & biomolecular chemistry (2011-12-24)
Hiroyuki Nakamura, Noriko Ueda, Hyun Seung Ban, Manabu Ueno, Shoji Tachikawa
ABSTRACT
The fluorescence-labeled closo-dodecaborane lipid (FL-SBL) was synthesized from (S)-(+)-1,2-isopropylideneglycerol as a chiral starting material. FL-SBL was readily accumulated into the PEGylated DSPC liposomes prepared from DSPC, CH, and DSPE-PEG-OMe by the post insertion protocol. The boron concentrations and the fluorescent intensities of the FL-SBL-labeled DSPC liposomes increased with the increase of the additive FL-SBL, and the maximum emission wavelength of the liposomes appeared at 531 nm. A preliminary in vivo imaging study of tumor-bearing mice revealed that the FL-SBL-labeled DSPC liposomes were delivered to the tumor tissue but not distributed to hypoxic regions.
Sigma-Aldrich
DL-α-Phosphatidylcholine, distearoyl, ≥98%
Sigma-Aldrich
1,2-Distearoyl-sn-glycero-3-phosphocholine, ≥99%