- Replacing phenyl ring with thiophene: an approach to longer wavelength aza-dipyrromethene boron difluoride (Aza-BODIPY) dyes.
Replacing phenyl ring with thiophene: an approach to longer wavelength aza-dipyrromethene boron difluoride (Aza-BODIPY) dyes.
The Journal of organic chemistry (2011-11-25)
Xinfu Zhang, Haibo Yu, Yi Xiao
PMID22111977
ABSTRACT
In the orignial 1,3,5,7-tetraphenyl aza-BODIPY, replacing the phenyl rings with thiophene achieved significant bathochromic shifts. One of the target molecules, DPDTAB, emitting strong NIR fluorescence with a quantum yield of 0.46 in acetonitrile, is a very competitive NIR fluorophore.
MATERIALS