Merck
  • Synthesis and field test of three candidates for soybean pod borer's sex pheromone.
Synthesis and field test of three candidates for soybean pod borer's sex pheromone.
Natural product communications (2011-09-29)
Tao Zhang, Juntao Feng, Chonglin Cai, Xing Zhang
ABSTRACT
Three candidates for the soybean pod borer's sex pheromone, dodec-10-en-1-yl acetate (E:Z = 95:5) (9a), (E, E)-dodeca-8, 10-dien-1-yl acetate (9b) and (E)-dodec-8-en-1-yl acetate (9c), were synthesized through the coupling reaction between Grignard reagents and acetates catalyzed by Li2CuCl4. Furthermore, the compounds 9a, 9b, and 9c, when tested in the field, showed that dodec-10-en-1-yl acetate (E:Z = 95:5) (9a) has promise as a lure for male soybean pod borer.
Product Description
Supelco
1-Dodecanol, Selectophore, ≥98.0%
Sigma-Aldrich
Lauryl alcohol, ≥98%, FG
Sigma-Aldrich
1-Dodecanol, ACS reagent, ≥98.0%
Sigma-Aldrich
1-Dodecanol, reagent grade, 98%