Merck
  • Application of an unusual ninhydrin-based reaction for the indirect chiral resolution of D,L-penicillamine.
Application of an unusual ninhydrin-based reaction for the indirect chiral resolution of D,L-penicillamine.
Talanta (2011-08-30)
Salvatore Sotgia, Angelo Zinellu, Gerard Aime Pinna, Luca Deiana, Ciriaco Carru
ABSTRACT
An unusual reaction involving ninhydrin and aminothiols was exploited to set an indirect method for the chiral recognition of stereoisomers of penicillamine. Separation of diastereoisomers was achieved on a C18 column in isocratic mode by using a mixture of propionic acid (pH 3.0)/acetonitrile/water (10:10:80, v/v/v) as a mobile phase. Diastereoisomers were detected by a fluorescence detector in fairly short times (about 7 min) and with a good resolution. The lowest detectable amount of toxic isomer of penicillamine (l-enantiomer) in samples of the d-enantiomer, was around 0.01%. The method was also suitable for the indirect chiral recognition of other aminothiols such as cysteine and cysteinylglycine.
Sigma-Aldrich
Ninhydrin, suitable for amino acid detection
Sigma-Aldrich
Ninhydrin, ACS reagent