Merck
  • Immobilization of MP-11 into a mesoporous metal-organic framework, MP-11@mesoMOF: a new platform for enzymatic catalysis.
Immobilization of MP-11 into a mesoporous metal-organic framework, MP-11@mesoMOF: a new platform for enzymatic catalysis.
Journal of the American Chemical Society (2011-06-21)
Vasiliki Lykourinou, Yao Chen, Xi-Sen Wang, Le Meng, Tran Hoang, Li-June Ming, Ronald L Musselman, Shengqian Ma
ABSTRACT
Microperoxidase-11 has for the first time been successfully immobilized into a mesoporous metal-organic framework (MOF) consisting of nanoscopic cages and it demonstrates superior enzymatic catalysis performances compared to its mesoporous silica counterpart.
Sigma-Aldrich
3,5-Di-tert-butylcatechol, 98%