Merck
  • Carotene and novel apocarotenoid concentrations in orange-fleshed Cucumis melo melons: determinations of β-carotene bioaccessibility and bioavailability.
Carotene and novel apocarotenoid concentrations in orange-fleshed Cucumis melo melons: determinations of β-carotene bioaccessibility and bioavailability.
Journal of agricultural and food chemistry (2011-03-23)
Matthew K Fleshman, Gene E Lester, Ken M Riedl, Rachel E Kopec, Sureshbabu Narayanasamy, Robert W Curley, Steven J Schwartz, Earl H Harrison
ABSTRACT
Muskmelons, both cantaloupe (Cucumis melo Reticulatus Group) and orange-fleshed honeydew (C. melo Inodorus Group), a cross between orange-fleshed cantaloupe and green-fleshed honeydew, are excellent sources of β-carotene. Although β-carotene from melon is an important dietary antioxidant and precursor of vitamin A, its bioaccessibility/bioavailability is unknown. We compared β-carotene concentrations from previously frozen orange-fleshed honeydew and cantaloupe melons grown under the same glasshouse conditions, and from freshly harvested field-grown, orange-fleshed honeydew melon to determine β-carotene bioaccessibility/bioavailability, concentrations of novel β-apocarotenals, and chromoplast structure of orange-fleshed honeydew melon. β-Carotene and β-apocarotenal concentrations were determined by HPLC and/or HPLC-MS, β-carotene bioaccessibility/bioavailability was determined by in vitro digestion and Caco-2 cell uptake, and chromoplast structure was determined by electron microscopy. The average β-carotene concentrations (μg/g dry weight) for the orange-fleshed honeydew and cantaloupe were 242.8 and 176.3 respectively. The average dry weights per gram of wet weight of orange-fleshed honeydew and cantaloupe were 0.094 g and 0.071 g, respectively. The bioaccessibility of field-grown orange-fleshed honeydew melons was determined to be 3.2 ± 0.3%, bioavailability in Caco-2 cells was about 11%, and chromoplast structure from orange-fleshed honeydew melons was globular (as opposed to crystalline) in nature. We detected β-apo-8'-, β-apo-10', β-apo-12'-, and β-apo-14'-carotenals and β-apo-13-carotenone in orange-fleshed melons (at a level of 1-2% of total β-carotene). Orange-fleshed honeydew melon fruit had higher amounts of β-carotene than cantaloupe. The bioaccessibility/bioavailability of β-carotene from orange-fleshed melons was comparable to that from carrot (Daucus carota).
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ≥96.0% (UV)
Sigma-Aldrich
trans-β-Apo-8′-carotenal, ~20% apocarotenal basis (UV-vis), suspension (oily)