Merck
Fire extinguisher: an imminent threat or an eminent danger?
The American journal of emergency medicine (2011-03-18)
Subramanian Senthilkumaran, Ramachandran Meenakshisundaram, Namasivayam Balamurgan, Krishnanswamy SathyaPrabhu, V Karthikeyan, Ponniah Thirumalaikolundusubramanian
ABSTRACT
Acute phosphate toxicity following rectal administration of phosphate-containing preparations was documented earlier. However, acute phosphate toxicity due to inhalation of monoammonium phosphate (MAP) is rarely reported. The present report describes the systemic toxicity following intentional inhalation of MAP, a dry chemical powder present in fire extinguishers, in a 25-year-old healthy man with normal renal function resulting in hyperphosphatemia (11.0 mg/dL), hypocalcemia (ionized calcium, 1.18 mg/dL; total calcium, 3.6 mg/dL), hypomagnesemia (1.2 mg/dL), seizures, and 4 episodes of pulseless polymorphic ventricular tachycardia requiring defibrillation. In view of the easy accessibility of fire extinguishers and its likely use for suicidal or homicidal purposes, physicians and practitioners working in emergency departments should be aware of this entity. Early recognition, removal of phosphate by hemodialysis, and supportive care have prevented this case from acute phosphate nephropathy and further complications.
Product Description
Supelco
Ammonium phosphate monobasic, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Sigma-Aldrich
Ammonium phosphate monobasic, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Ammonium dihydrogenphosphate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium phosphate dibasic, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium phosphate dibasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, suitable for plant cell culture
Sigma-Aldrich
Ammonium phosphate dibasic, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98% (alkalimetric)
Sigma-Aldrich
Ammonium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate monobasic, ACS reagent, ≥98%
Sigma-Aldrich
Ammonium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Ammonium phosphate monobasic, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium phosphate monobasic, ReagentPlus®, ≥98.5%