Merck
  • A multipathogen selective enrichment broth for simultaneous growth of Salmonella spp., Vibrio parahaemolyticus, and Vibrio cholerae.
A multipathogen selective enrichment broth for simultaneous growth of Salmonella spp., Vibrio parahaemolyticus, and Vibrio cholerae.
The Journal of general and applied microbiology (2011-02-02)
Xing-Long Xiao, Yi-Juan Li, Yi-Ying Qin, Yi-Gang Yu, Hui Wu
ABSTRACT
A selective enrichment broth (SVV) was formulated to allow concurrent growth of Salmonella spp., V. parahaemolyticus, and V. cholerae. Potassium tellurite and sodium citrate were added as the inhibitors, while glucose, mannitol, anhydrous sodium sulfite and sodium pyruvate were employed as the growth-promoters. When mixed in equal or varied proportions, the target pathogens in SVV had a great accumulation (10(5)-10(8) CFU/ml) and effectively inhibited the growth of competitive microflora. In the artificially contaminated samples, a high recovery of these 3 target pathogens was obtained in SVV. Finally, Salmonella spp., V. parahaemolyticus, and V. cholerae were detected from 608 suspicious food samples by SVV with real-time PCR, and no false-positive or -negative results were reported. In summary, SVV has been shown to be a suitable broth for the simultaneous detection of the 3 pathogens by multipathogen detection on a single-assay platform.
Product Description
Millipore
Potassium tellurite solution, 1% in H2O, suitable for microbiology