Merck
  • Listeria monocytogenes exploits cystic fibrosis transmembrane conductance regulator (CFTR) to escape the phagosome.
Listeria monocytogenes exploits cystic fibrosis transmembrane conductance regulator (CFTR) to escape the phagosome.
Proceedings of the National Academy of Sciences of the United States of America (2011-01-12)
Andrea L Radtke, Kelsi L Anderson, Michael J Davis, Matthew J DiMagno, Joel A Swanson, Mary X O'Riordan
ABSTRACT
Virulence of the intracellular pathogen Listeria monocytogenes (Listeria) requires escape from the phagosome into the host cytosol, where the bacteria replicate. Phagosomal escape is a multistep process characterized by perforation, which is dependent on the pore-forming toxin listeriolysin O (LLO), followed by rupture. The contribution of host factors to Listeria phagosomal escape is incompletely defined. Here we show that the cystic fibrosis transmembrane conductance regulator (CFTR) facilitates Listeria cytosolic entry. CFTR inhibition or mutation suppressed Listeria vacuolar escape in culture, and inhibition of CFTR in wild-type mice before oral inoculation of Listeria markedly decreased systemic infection. We provide evidence that high chloride concentrations may facilitate Listeria vacuolar escape by enhancing LLO oligomerization and lytic activity. We propose that CFTR transiently increases phagosomal chloride concentration after infection, potentiating LLO pore formation and vacuole lysis. Our studies suggest that Listeria exploits mechanisms of cellular ion homeostasis to escape the phagosome and emphasize host ion-channel function as a key parameter of bacterial virulence.
Sigma-Aldrich
N-Phenylanthranilic acid, 98%