Merck
  • Colorimetric assay for cyanide and cyanogenic glycoside using polysorbate 40-stabilized gold nanoparticles.
Colorimetric assay for cyanide and cyanogenic glycoside using polysorbate 40-stabilized gold nanoparticles.
Chemical communications (Cambridge, England) (2011-01-11)
Cheng-Yu Liu, Wei-Lung Tseng
ABSTRACT
This study describes a simple method for the selective and sensitive detection of cyanide and endogenous biological cyanide using polysorbate 40-stabilized gold nanoparticles.
Sigma-Aldrich
TWEEN® 40, viscous liquid
Sigma-Aldrich
ECO TWEEN® 40, viscous liquid