Merck
  • Pathogenicity of exonic indels in fused in sarcoma in amyotrophic lateral sclerosis.
Pathogenicity of exonic indels in fused in sarcoma in amyotrophic lateral sclerosis.
Neurobiology of aging (2010-11-16)
Nicola J Rutherford, Nicole A Finch, Mariely DeJesus-Hernandez, Richard J P Crook, Catherine Lomen-Hoerth, Zbigniew K Wszolek, Ryan J Uitti, Neill R Graff-Radford, Rosa Rademakers
ABSTRACT
Insertion and deletion variants (indels) within poly glycine tracts of fused in sarcoma (FUS) were initially reported as causative of disease in amyotrophic lateral sclerosis (ALS). Subsequent studies identified similar indels in controls and suggested that these indels may confer susceptibility to ALS. We aimed to elucidate the role of previously published and novel exonic indels in FUS in an extensive cohort of 630 ALS patients and 1063 controls. We detected indels in FUS exons 5, 6, 12, and 14 with similar frequencies in patients (0.95%) and controls (0.75%). Exonic indels in poly glycine tracts were also observed with similar frequencies. The largest indel (p.Gly138_Tyr143del) was observed in 1 control. In 1 patient, a 3 base pair deletion in exon 14 (p.Gly475del) was identified, however in vitro studies did not reveal abnormal localization of p.Gly475del mutant FUS. These findings suggest that not all exonic indels in FUS cause disease.
Sigma-Aldrich
Polyglycine, mol wt 500-5,000