Merck
  • Improved synthesis of (E)-12-nitrooctadec-12-enoic acid, a potent PPARγ activator. Development of a "buffer-free" enzymatic method for hydrolysis of methyl esters.
Improved synthesis of (E)-12-nitrooctadec-12-enoic acid, a potent PPARγ activator. Development of a "buffer-free" enzymatic method for hydrolysis of methyl esters.
The Journal of organic chemistry (2010-11-11)
Giuseppe Zanoni, Matteo Valli, Lyamin Bendjeddou, Alessio Porta, Paolo Bruno, Giovanni Vidari
ABSTRACT
Endogenous nitro-fatty acids, acting as partial agonist of PPARγ, are able to lower the insulin and glucose levels without the side effects associated with common antidiabetic drugs. (E)-12-Nitrooctadec-12-enoic acid, a potent activator of this peroxisome receptor, was synthesized in a very efficient sequence via a Henry-retro-Claisen ring fragmentation, followed by a novel enzymatic cleavage of methyl esters. The latter method was then applied in the last step of the synthesis of a few labile natural products, such as prostaglandins, isoprostanes, and phytoprostanes.
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%
Supelco
Trifluoroacetic anhydride, for GC derivatization, LiChropur, ≥99.0% (GC)