- Enantioselective synthesis of (R)-bufuralol via dynamic kinetic resolution in the key step.
Enantioselective synthesis of (R)-bufuralol via dynamic kinetic resolution in the key step.
The Journal of organic chemistry (2010-06-08)
Eric V Johnston, Krisztián Bogár, Jan-E Bäckvall
PMID20524620
ABSTRACT
An enantioselective synthesis of (R)-bufuralol via a ruthenium- and enzyme-catalyzed dynamic kinetic resolution (DKR) has been achieved. The synthesis starts from readily available 2-ethylphenol and provides (R)-bufuralol in high ee and a good overall yield of 31%.
MATERIALS