Merck
  • Azulenocyanine: a new family of phthalocyanines with intense near-IR absorption.
Azulenocyanine: a new family of phthalocyanines with intense near-IR absorption.
Journal of the American Chemical Society (2010-05-27)
Atsuya Muranaka, Mitsuhiro Yonehara, Masanobu Uchiyama
ABSTRACT
Azulenocyanine, having four azulene units fused to a tetraazaporphyrin skeleton, is a structural isomer of naphthalocyanine. We synthesized the first example of an azulenocyanine from 1,3-di-tert-butyl-5,6-dicyanoazulene. The macrocycle exhibits broad absorption over the visible and near-IR regions far beyond 1000 nm. Theoretical calculations and electrochemical experiments show the LUMO energy level is drastically lowered. Azulenocyanine represents a new type of oxidation-stable near-IR dye.
Sigma-Aldrich
Azulene, 99%