Merck
  • Noncrystalline NiPB nanotubes for hydrogenation of p-chloronitrobenzene.
Noncrystalline NiPB nanotubes for hydrogenation of p-chloronitrobenzene.
Chemical communications (Cambridge, England) (2010-03-18)
Min Mo, Ling Han, Jiangang Lv, Yan Zhu, Luming Peng, Xuefeng Guo, Weiping Ding
ABSTRACT
High quality noncrystalline NiPB nanotubes were synthesized and exhibited high efficiency for the catalytic hydrogenation of p-chloronitrobenzene due to the characteristic confinement effect of the nanotubes.
Sigma-Aldrich
1-Chloro-4-nitrobenzene, 99%