Merck
  • Preparation and characteristics of protein molecularly imprinted membranes on the surface of multiwalled carbon nanotubes.
Preparation and characteristics of protein molecularly imprinted membranes on the surface of multiwalled carbon nanotubes.
Talanta (2010-03-02)
Maosheng Zhang, Jiarong Huang, Ping Yu, Xi Chen
ABSTRACT
A novel protein molecularly imprinted membrane (PMIM) was synthesized on the surface of multiwalled carbon nanotubes (MWNTs) through a surface molecular imprinting technique by using bovine serum albumin (BSA) as the template molecule, acrylamide (AAm) as the functional monomer, N,N'-methylenebisacrylamide (NNMBA) as the cross-linker and ammonium persulphate [(NH(4))(2)S(2)O(8)] as the initiator. The amounts of raw materials were optimized in this paper and the suitable amount is 0.1g of CNTs, 0.02 g of BSA, 0.24 g of AAm, 0.1g of MBA and 0.025 mg of (NH(4))(2)S(2)O(8). The selective recognition ability of PMIM/MWNTs was evaluated using adsorption experiments. Maximum adsorption capacity was 5.53 microg/mg PMIM/MWNTs and a saturation value was achieved at a BSA concentration of 0.2mg/mL. The selectivity adsorption experiments showed that the PMIM/MWNTs also had higher adsorption capacities for BSA than for such molecules, as HSA, HB, pepsin and HRP. The PMIM/MWNTs displayed a 2.6-fold increase in affinity to BSA compared to the nPMIM/MWNTs. The PMIM/MWNTs, on the other hand, did not exhibit any significant change in affinity to other molecules compared to the nPMIM/MWNTs. In the experiment, the characteristics of the PMIM/MWNTs were analyzed using infrared spectroscopy and their configurations were observed using scanning electron microscopy.
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, for molecular biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O