Merck
  • Application of hydrophilic interaction chromatography retention coefficients for predicting peptide elution with TFA and methanesulfonic acid ion-pairing reagents.
Application of hydrophilic interaction chromatography retention coefficients for predicting peptide elution with TFA and methanesulfonic acid ion-pairing reagents.
Journal of separation science (2010-01-21)
Chad E Wujcik, Joseph Tweed, Eugene P Kadar
ABSTRACT
Hydrophilic retention coefficients for 17 peptides were calculated based on retention coefficients previously published for TSKgel silica-60 and were compared with the experimental elution profile on a Waters Atlantis HILIC silica column using TFA and methanesulfonic acid (MSA) as ion-pairing reagents. Relative peptide retention could be accurately determined with both counter-ions. Peptide retention and chromatographic behavior were influenced by the percent acid modifier used with increases in both retention and peak symmetry observed at increasing modifier concentrations. The enhancement of net peptide polarity through MSA pairing shifted retention out by nearly five-fold for the earliest eluting peptide, compared with TFA. Despite improvements in retention and efficiency (N(eff)) for MSA over TFA, a consistent reduction in calculated selectivity (alpha) was observed. This result is believed to be attributed to the stronger polar contribution of MSA masking and diminishing the underlying influence of the amino acid residues of each associated peptide. Finally, post-column infusion of propionic acid and acetic acid was evaluated for their potential to recover signal intensity for TFA and MSA counter-ions for LC-ESI-MS applications. Acetic acid generally yielded more substantial signal improvements over propionic acid on the TFA system while minimal benefits and some further reductions were noted with MSA.
Sigma-Aldrich
Potassium methanesulfonate, ≥98.0% (dry substance, T)
Sigma-Aldrich
Sodium methanesulfonate, 98%
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Silver methanesulfonate
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC