Merck
  • Quantification of (15)N-nitrate in urine with gas chromatography combustion isotope ratio mass spectrometry to estimate endogenous NO production.
Quantification of (15)N-nitrate in urine with gas chromatography combustion isotope ratio mass spectrometry to estimate endogenous NO production.
Analytical chemistry (2009-12-17)
Els Houben, Henrike M Hamer, Anja Luypaerts, Vicky De Preter, Pieter Evenepoel, Paul Rutgeerts, Kristin Verbeke
ABSTRACT
The use of stable isotope labeled substrates and subsequent analysis of urinary nitrate, forms a noninvasive test for evaluation of the in vivo NO metabolism. The present paper describes a new method for simultaneous quantification of (15)N-nitrate and total nitrate with gas chromatography combustion isotope ratio mass spectrometry (GC-C-IRMS). Nitrate, isolated from urine with a nitrate selective resin, was reduced to nitrite using copperized cadmium. Subsequently, Sudan I was formed by diazotation. Sudan II was added as internal standard, and both molecules were analyzed with GC-C-IRMS as tert-butyldimethylsilyl derivatives. The accuracy was determined during a recovery study of two different known nitrate concentrations and two (15)N-enrichments. A recovery of 101.6% and 103.9% for total nitrate and 107.6% and 91.2% for (15)N-nitrate was obtained, respectively. The validated method was applied on complete 72 h urine collections after intravenous administration of (15)N-nitrate and (15)N-arginine in humans. On average, 51.8% (47.0-71.0%) of administered (15)N-nitrate was excreted, while 0.68% (0.44-1.17%) of (15)N-arginine was metabolized to nitrate. In conclusion, this method can be used for accurate simultaneous determination of (15)N-nitrate and total nitrate concentrations in urine and can be applied in clinical studies for noninvasive evaluation of NO metabolism in vivo.
Product Description
Supelco
Sudan II, analytical standard
Sigma-Aldrich
Sudan II, Dye content 90 %