Merck
  • Expression of cyclooxygenase-1/-2, microsomal prostaglandin-E synthase-1 and E-prostanoid receptor 2 and regulation of inflammatory mediators by PGE(2) in the amoeboid microglia in hypoxic postnatal rats and murine BV-2 cells.
Expression of cyclooxygenase-1/-2, microsomal prostaglandin-E synthase-1 and E-prostanoid receptor 2 and regulation of inflammatory mediators by PGE(2) in the amoeboid microglia in hypoxic postnatal rats and murine BV-2 cells.
Neuroscience (2009-08-29)
P Li, J Lu, C Kaur, V Sivakumar, K L Tan, E A Ling
ABSTRACT
This study aimed to investigate the effect of hypoxia on the expression of cyclooxygenase-1 (COX-1), cyclooxygenase-2 (COX-2), microsomal prostaglandin-E synthase (mPGES-1), E-prostanoid receptor 2 (EP2) in microglia; and the roles of EP2-cyclic adenosine monophosphate (cAMP) signaling pathway in the prostaglandin E(2) (PGE(2)) regulation of inflammatory mediators released by hypoxic BV-2 cells. Immunoexpression of COX-1, COX-2, mPGES-1 and EP2 was localized in the amoeboid microglial cells (AMC), a nascent brain macrophage in the developing brain, as confirmed by double labeling with OX-42 and lectin, specific markers of microglia. AMC emitted a more intense immunofluorescence in hypoxic rats when compared with the matching controls. In postnatal rats subjected to hypoxia, mRNA and protein expression levels of COX-1, COX-2 and mPGES-1 along with tumor necrosis factor-alpha (TNF-alpha), interleukin-1beta (IL-1beta), inducible nitric-oxide synthase (iNOS) and PGE(2) product in the callosal tissue were significantly increased. The results were shared in the BV-2 cells except for COX-1 mRNA and protein whose levels remained unaltered. Interestingly, treatment with EP2 antagonist AH-6809 resulted in suppression of hypoxia induced EP2, IL-1beta and iNOS mRNA and protein expression, TNF-alpha protein expression and intracellular cAMP level in BV-2 cells. It is suggested that PGE(2) may regulate above inflammatory mediators in the activated microglia via EP2-cAMP signaling pathway in hypoxic conditions.
Sigma-Aldrich
AH6809, ≥98%, crystalline solid or supercooled liquid