Merck
Sources and prevalence of pentachlorobenzene in the environment.
Chemosphere (2009-03-21)
Robert E Bailey, Dolf van Wijk, Paul C Thomas
ABSTRACT
There are no longer any large scale uses of pentachlorobenzene (PeCB). Current emissions of PeCB to the environment are estimated to be about 121000kgy(-1), based on published information. The largest sources appear to be combustion of solid wastes, 32000kgy(-1), biomass burning, 45000kgy(-1) with degradation of an agricultural fungicide, quintozene, contributing 26000kgy(-1) and industrial releases less important. PeCB has been measured in many environmental media over the past 35 years. Low but detectable concentrations of PeCB have been reported in the atmosphere, sediments and biota in remote areas of the world. Calculations using a global distribution model are consistent with the estimate of approximately 100000kgy(-1) global PeCB emissions. Concentrations of PeCB in the environment have declined with a 90% decrease of PeCB concentrations in herring gull eggs from Lake Superior, Canada since 1979.
Sigma-Aldrich
Pentachlorobenzene, 96%
Supelco
Pentachlorobenzene, PESTANAL®, analytical standard