Merck
Microchip assays for screening monoclonal antibody product quality.
Electrophoresis (2009-01-09)
Xiaoyu Chen, Kaiyan Tang, Maximilian Lee, Gregory C Flynn
ABSTRACT
Microchip CE-SDS was evaluated as a high-throughput alternative to conventional CE-SDS for monitoring monoclonal antibody protein quality. A commercial instrument (LabChip) 90) was used to separate dodecyl sulfate coated proteins through a sieving polymer based on the proteins' sizes. Under reducing conditions, the microchip CE-SDS separation was similar to that of conventional CE-SDS, providing reasonable resolution of the non-glycosylated and the glycosylated heavy chains. The fluorescence detection on LabChip 90 using non-covalent fluorescent labeling method was about as sensitive as the 220 nm UV detection used in a conventional CE instrument. A simple glycan typing assay was developed for the reducing microchip CE-SDS format. Antibodies, either pure or in crude cell culture media are treated with Endoglycosidase H, which specifically cleaves the hybrid and high mannose type glycans. A heavy chain migration shift on reducing CE-SDS resulting from the loss of glycan is used to measure the level of high mannose/hybrid type glycans as a percentage of the total glycans. Microchip CE-SDS, under both non-reducing and reducing conditions, can be used in a variety of antibody product screening assays. The microchip analyses provide sufficient resolution and sensitivity for this purpose but on a time scale approximately 70 times faster (41 s versus 50 min per sample) than conventional CE separation under typical operational conditions.
Sigma-Aldrich
Lithium dodecyl sulfate, BioXtra, ≥98.5% (GC)
Sigma-Aldrich
Lithium dodecyl sulfate, BioReagent, for molecular biology, suitable for electrophoresis
Sigma-Aldrich
Lithium dodecyl sulfate, ≥98.5% (GC)
Sigma-Aldrich
Ammonium lauryl sulfate solution, ~30% in H2O (T)
Sigma-Aldrich
Ammonium lauryl sulfate solution, ~30% in H2O