Merck
A glimpse of the inner workings of the templated site.
Chemical communications (Cambridge, England) (2008-12-23)
Christopher J Allender, Oliver K Castell, Philip R Davies, Steven Fiddy, Jimmy Hedin-Dahlström, Michael Stockenhuber
ABSTRACT
This is the first direct experimental probe, using EXAFS, of the active site within molecularly imprinted polymers and paves the way to a more detailed understanding of the inner workings of molecular imprinting.
Sigma-Aldrich
1,3-Diphenyl-1,3-propanedione, 98%
Sigma-Aldrich
Polystyrene, average Mw ~280,000 by GPC
Sigma-Aldrich
Polystyrene, average Mw ~192,000
Sigma-Aldrich
Polystyrene, average Mw ~350,000, average Mn ~170,000
Sigma-Aldrich
Polystyrene, average Mw ~400,000 by GPC
Sigma-Aldrich
Polystyrene, average Mw 35,000