Merck
  • Aluminum-catalyzed asymmetric alkylations of pyridyl-substituted alkynyl ketones with dialkylzinc reagents.
Aluminum-catalyzed asymmetric alkylations of pyridyl-substituted alkynyl ketones with dialkylzinc reagents.
Journal of the American Chemical Society (2008-07-01)
Donna K Friel, Marc L Snapper, Amir H Hoveyda
ABSTRACT
Alkylations of pyridyl-substituted ynones with Et2Zn and Me2Zn, promoted by amino acid-based chiral ligands in the presence of Al-based alkoxides, afford tertiary propargyl alcohols efficiently in 57% to >98% ee. Two easily accessible chiral ligands are identified as optimal for reactions of the two dialkylzinc reagents. Catalytic alkylations with Et2Zn require a chiral ligand carrying two amino acid moieties (valine and phenylalanine) along with a p-trifluoromethylphenylamide C-terminus. In contrast, reactions with Me2Zn are most effectively promoted in the presence of a chiral ligand containing a single amino acid (benzyl cysteine), capped by an n-butylamide. Enantiomerically enriched tertiary alcohols bearing a pyridyl and an alkyne substituent can be functionalized in a variety of manners to furnish a wide range of difficult-to-access acyclic and heterocyclic structures; two noteworthy examples are Cu-catalyzed protocols for conversion of tertiary propargyl alcohols to enantiomerically enriched tetrasubstituted allenes and bicyclic amides that bear an N-substituted quaternary carbon stereogenic center. Mechanistic models that account for the trends and enantioselectivity levels are provided.
Sigma-Aldrich
Diethylzinc, ≥52 wt. % Zn basis
Sigma-Aldrich
Diethylzinc solution, 1.0 M in hexanes
Sigma-Aldrich
Diethylzinc solution, 15 wt. % in toluene
Sigma-Aldrich
Diethylzinc solution, 1.0 M in heptane