Merck
  • 4-ethylphenol and 4-ethylguaiacol in wines: estimating non-microbial sourced contributions and toxicological considerations.
4-ethylphenol and 4-ethylguaiacol in wines: estimating non-microbial sourced contributions and toxicological considerations.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2007-11-06)
Sierra Rayne, Nigel J Eggers
ABSTRACT
Analyses of commercially available wines suggested non-Brettanomyces sources of 4-ethylphenol and 4-ethylguaiacol. Grapes, enological additions, exposure to plastics, and oak-barrel aging were potential inputs considered. Investigations of whole grape bunch samples from two major red wine Vitis vinifera cultivars (L. cv. Cabernet Franc and Pinot Noir), a commercial mannoprotein additive, and three commercial enological tannin additions indicated they are not likely significant sources of these compounds. Studies on 15 commercial oak barrelled red wines from six Vitis vinifera cultivars (L. cv. Cabernet Franc, Cabernet Sauvignon, Dunkelfelder, Merlot, Pinot Meunier, and Pinot Noir), and a review of volatile phenol extraction from toasted oak wood, suggested that oak-aging may produce concentrations of up to 50 microg L(-1) 4-ethylphenol and 4-ethylguaiacol. Thus, following potential Brettanomyces-sourced aroma impacts in wine using 4-ethylphenol and/or 4-ethylguaiacol concentrations as proxies should only be considered reliable at analyte levels>100 microg L(-1). A review of worldwide 4-ethylphenol and 4-ethylguaiacol concentrations in wine, consumption patterns, and available toxicological data also suggested that levels of 4-ethylphenol being observed in wines worldwide do not warrant concerns about acute or long-term effects. While little is known about the toxicology of 4-ethylguaiacol, it is unlikely that elevated concentrations will pose any health-related concerns.
Sigma-Aldrich
4-Ethylguaiacol, natural, FG
Sigma-Aldrich
4-Ethylguaiacol, ≥98%, FCC, FG